ContactUsLinkComponent
Chloramphenicol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Chloramphenic ol in tissue, milk, honey, water, egg. 1 * 32 Tests
Catalog # ANTIA327190-32
Supplier:  ANTIBODIES.COM
New Product
Chloramphenicol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Chloramphenic ol in tissue, milk, honey, water, egg. 1 * 32 Tests
Catalog # ANTIA327190-32
Supplier:  ANTIBODIES.COM