Aflatoxin M1 Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Aflatoxin M1 in raw commingled cow’ s milk. 1 * 96 Tests
Catalog # ANTIA327179-96
Supplier: ANTIBODIES.COM