Rat Troponin I Fast Skeletal Muscle ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of rat Troponin I Fast Skeletal Muscle in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5456-48
Supplier: ANTIBODIES.COM