ContactUsLinkComponent
Human IL-36 gamma/IL-1F9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human IL-36 gamma/IL-1F9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA5223-48
Supplier:  ANTIBODIES.COM
New Product
Human IL-36 gamma/IL-1F9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human IL-36 gamma/IL-1F9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA5223-48
Supplier:  ANTIBODIES.COM