Beta Lactam + Tetracycline + Cephalexin + Ceftiofur Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualit ative determination of Beta Lactam + Tetracycline + Cephalexin + Ceftiofur in milk. 1 * 96 Tests
Catalog # ANTIA327184-96
Supplier: ANTIBODIES.COM