ContactUsLinkComponent
Mouse IL-36 gamma/IL-1F9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse IL-36 gamma/IL-1F9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA5704-48
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-36 gamma/IL-1F9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse IL-36 gamma/IL-1F9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA5704-48
Supplier:  ANTIBODIES.COM