ContactUsLinkComponent
Human IL-18R beta/IL-18RAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-18R beta/IL-18RAP in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5559-48
Supplier:  ANTIBODIES.COM
New Product
Human IL-18R beta/IL-18RAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-18R beta/IL-18RAP in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5559-48
Supplier:  ANTIBODIES.COM