Human SARS-CoV-2 Neutralization Antibody ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay designed for the in vitro quan titative determination of human SARS-CoV-2 Neutralization Antibody in serum and plasma. 1 * 96 Tests
ANTIA327005-96