ContactUsLinkComponent
Rat MIP-3 beta/CCL19 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat MIP-3 beta/CCL19 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 96 Tests
Catalog # ANTIA3461-96
Supplier:  ANTIBODIES.COM
New Product
Rat MIP-3 beta/CCL19 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat MIP-3 beta/CCL19 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 96 Tests
Catalog # ANTIA3461-96
Supplier:  ANTIBODIES.COM