Gentamicin + Kanamycin + Streptomycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determin ation of Gentamicin + Kanamycin + Streptomycin in raw commingled cow’ s milk. 1 * 96 Tests
Catalog # ANTIA327219-96
Supplier: ANTIBODIES.COM