Human IL-13 Receptor alpha 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of human IL-13 Receptor alpha 2 in serum, plasma, tissue homogenates, cell lysates, cell culture sup 1 * 96 Tests
ANTIA7868-96