Mouse alpha Smooth Muscle Actin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse alpha Smooth Muscle Actin in serum, plasma, tissue homogenates or other biological fluid 1 * 48 Tests
Catalog # ANTIA82339-48
Supplier: ANTIBODIES.COM