ContactUsLinkComponent
Human RCC1 and BTB Domain Containing Protein 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the i n vitro quantitative determination of human RCC1 and BTB Domain Containing Protein 2 in tissue homogenates or other b 1 * 48 Tests
Catalog # ANTIA7217-48
Supplier:  ANTIBODIES.COM
New Product
Human RCC1 and BTB Domain Containing Protein 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the i n vitro quantitative determination of human RCC1 and BTB Domain Containing Protein 2 in tissue homogenates or other b 1 * 48 Tests
Catalog # ANTIA7217-48
Supplier:  ANTIBODIES.COM