ContactUsLinkComponent
,,Conductivity Standard 100000 uS/cm at 25 C CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 12 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, 1 * 500 mL
Catalog # CMPLCS1P5S.L5
New Product
,,Conductivity Standard 100000 uS/cm at 25 C CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 12 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, 1 * 500 mL
Catalog # CMPLCS1P5S.L5