ContactUsLinkComponent
2 components: Zn 80g/l Cd 20g/l in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1.000 mL
Catalog # CMPLRM1180341L
New Product
2 components: Zn 80g/l Cd 20g/l in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1.000 mL
Catalog # CMPLRM1180341L