ContactUsLinkComponent
1 component: Trifluoroacetic Acid [CAS:76-05-1] 100ug/ml in Methanol / Water (1/1) CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1 mL
Catalog # CMPLRD1179201
New Product
1 component: Trifluoroacetic Acid [CAS:76-05-1] 100ug/ml in Methanol / Water (1/1) CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1 mL
Catalog # CMPLRD1179201