ContactUsLinkComponent
,,34 components: Al 100mg/l As 100mg/l B 100mg/l Ba 100mg/l Bi 100mg/l Ca 100mg/l Cd 100mg/l Co 100mg/l Cr 100mg/l Cu 100mg/l Fe 100mg/l In 100mg/l K 100mg/l Li 100mg/l Mg 100mg/l Mn 100mg/l Mo 100mg/l Na 100mg/l N 1 * 100 mL
Catalog # CMPLRM117322L1
New Product
,,34 components: Al 100mg/l As 100mg/l B 100mg/l Ba 100mg/l Bi 100mg/l Ca 100mg/l Cd 100mg/l Co 100mg/l Cr 100mg/l Cu 100mg/l Fe 100mg/l In 100mg/l K 100mg/l Li 100mg/l Mg 100mg/l Mn 100mg/l Mo 100mg/l Na 100mg/l N 1 * 100 mL
Catalog # CMPLRM117322L1