DINO-LITE MICROSCOPE AM8917MT8A 1 * 1 items
Catalog # IDCPAM8917MT8A
Supplier: Dino-Lite / IDCP B.V.
New Product
DINO-LITE MICROSCOPE AM8917MT8A 1 * 1 items
Catalog # IDCPAM8917MT8A
Supplier: Dino-Lite / IDCP B.V.
New Product
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
VWR® Autoclavable Biohazard Bags, Polypropylene, Orange
Supplier: VWR Collection
VWR® Reagent Reservoirs
Supplier: VWR Collection
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Nitrile gloves, Kimtech™ Blue Nitrile™
Supplier: KIMBERLY CLARK