ContactUsLinkComponent
EZ4 W without eyepieces Microscope Functions: 4.4:1 Zoom Stereo Microscope without eyepieces with built- in 5 MPixel HD digital camera and 0.8x-3.5x zoom range, (customer can choose from 10x and 16x high eyepoint eyepieces) Leica exclusive 50-75 mm 1 * 1 Each
Catalog # LEIM10450948
Supplier:  LEICA MICROSYSTEMS
New Product
EZ4 W without eyepieces Microscope Functions: 4.4:1 Zoom Stereo Microscope without eyepieces with built- in 5 MPixel HD digital camera and 0.8x-3.5x zoom range, (customer can choose from 10x and 16x high eyepoint eyepieces) Leica exclusive 50-75 mm 1 * 1 Each
Catalog # LEIM10450948
Supplier:  LEICA MICROSYSTEMS