14 components: Ca 5mg/l Mo 10mg/l K 10mg/l Zn 10mg/l Mn 10mg/l Ni 10mg/l Na 10mg/l Fe 10mg/l Cu 10mg/l Al 10mg/l Ti 10mg/l Si 10mg/l P 20mg/l Mg 5mg/l in HNO3 5% 1 * 1 L
Catalog # CMPLRM1129001L
Supplier: CUSTOM MADE CHEMICALS LAB