Orders LinkContactUsLinkComponent
Objectives
Objectives
Universal plan extended apochromat objective, 0,18 mm
Catalog # OLYMN5742100
Supplier:   OLYMPUS OPTICAL
Objectives
Catalog # OLYMN5742100
Supplier:  OLYMPUS OPTICAL
Objectives
Objectives
Universal plan extended apochromat objective, 0,18 mm
Price
659,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    40X
  • Description:
    Universal plan extended apochromat objective
  • Seq:
    05
  • Working distance:
    0,18 mm