ContactUsLinkComponent
27 components: 10mg/l each of Al Sb As Ba Be Cd Ca Co Cr Fe P Mg Mn Hg Mo Ni Pb K Cu Se Na Sn Tl Te U V Zn in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laborato 1 * 50 mL
Catalog # CMPLRM110743L05
New Product
27 components: 10mg/l each of Al Sb As Ba Be Cd Ca Co Cr Fe P Mg Mn Hg Mo Ni Pb K Cu Se Na Sn Tl Te U V Zn in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laborato 1 * 50 mL
Catalog # CMPLRM110743L05