JERRICAN HDPE 5L CAP WHITE NON TAMP/EV 1 * 180 items
Catalog # VWRL080504
Supplier: LAB3 OWN PRODUCTION
New Product
JERRICAN HDPE 5L CAP WHITE NON TAMP/EV 1 * 180 items
Catalog # VWRL080504
Supplier: LAB3 OWN PRODUCTION
New Product
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Labels, Cryo
Supplier: VWR Collection
VWR® Beakers, Disposable
Supplier: VWR Collection
VWR® Cooling Bricks
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Pipette Tips, Standard
Supplier: VWR Collection
TAPE MICROPORE 9MX25MM BLUE DISPENSER 1 * 12 items
Supplier: 3M MEDICAL
Sterilin™ Quickstart Universal Containers, PS, 30 ml
Supplier: Sterilin
Dialysis tubing Spectra/Por® 5
Supplier: Spectrum Laboratories
VWR® Reagent Reservoirs
Supplier: VWR Collection
Coverplast Barrier Plasters Washproof 7.2 x 2.2cm 1 * 100 items
Supplier: SAFETY FIRST AID GROUP