Label Sheets, Core2Write
Core2Write label, sterile, irradiated, White, 76,2×25,4 mm, 18 labels/sheet, 500 sheets/case
VELTC2WR3X1SHEET02
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures.
- Can be applied to sub-zero surfaces
- Perforation between labels is available
All labels can be customised or left blank.
: Also available in sheets (sold separately).
- Colour:White
- Description:Core2Write label, sterile, irradiated
- Dimensions:76,2×25,4 mm
- Seq:03
- Packed:18 labels/sheet, 500 sheets/case