Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
8 components: Ni 100mg/l Cu 100mg/l Co 100mg/l Pb 40mg/l As 40mg/l Zn 40mg/l Cd 20mg/l Hg 20mg/l in HNO3 5% CRM, ISO 17034 and ISO 17025 1 * 200 mL
ACSDRM107895L2
:
CPACHEM
New Product
8 components: Ni 100mg/l Cu 100mg/l Co 100mg/l Pb 40mg/l As 40mg/l Zn 40mg/l Cd 20mg/l Hg 20mg/l in HNO3 5% CRM, ISO 17034 and ISO 17025 1 * 200 mL
ACSDRM107895L2
:
CPACHEM
New Product
Frequently Bought Together
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
pH buffers, ready to use
: WTW
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals