Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1 COMPONENT: HG 0.075MG/L IN HCL 5% 1 * 250 mL
Catalog # CMPLRM076382L25
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Expand image
1 COMPONENT: HG 0.075MG/L IN HCL 5% 1 * 250 mL
Catalog # CMPLRM076382L25
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Sodium hypochlorite (14% Cl₂) in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach