6 components: K 20000mg/l Ca 2500mg/l Al 20mg/l Cu 20mg/l Sr 20mg/l Ba 4mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 5 Bottle
Catalog # CMPLRM102911L1.5
Supplier: CUSTOM MADE CHEMICALS LAB