PAH SOLUTION 16 COMPONENTS 100MG/L EACH 1 * 100 mL
Catalog # CMPLRM022905L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
PAH SOLUTION 16 COMPONENTS 100MG/L EACH 1 * 100 mL
Catalog # CMPLRM022905L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
2-Methylbutane for HPLC
Supplier: Thermo Fisher Scientific
Pyridine 99+%, extra pure
Supplier: Thermo Fisher Scientific
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
ETHYLENE GLYCOL ≥98 TECHNICAL 1 * 5 L
Supplier: Roth Carl
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, anhydrous ≥99.8% unstabilised
Supplier: Thermo Fisher Scientific
Water for analysis
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific