IQ/OQ documentation for: POLYMIX PX-MFC 90 1 * 1 items
Catalog # KINEDIV
Supplier: KINEMATICA
New Product
IQ/OQ documentation for: POLYMIX PX-MFC 90 1 * 1 items
Catalog # KINEDIV
Supplier: KINEMATICA
New Product
Frequently Bought Together
NEW POWER SUPPLY FOR THE AIR SAMPLER MD8 1 * 1 items
Supplier: Sartorius
HOUSING CLIP REAR FOR HANDYSTEP S 1 * 1 items
Supplier: Brand
CIRCLIP PLIERS SIZE S1 1 * 1 ST
Supplier: IKA
Assembled pump NaOH 230V 1 * 1 items
Supplier: VELP SCIENTIFIC
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
SERVICE KIT PCRW01 BACK PANEL BE 1 * 1 items
Supplier: VWR Collection
CRYOPLUS PCB MINI T B/PASS PRO KT 1 * 1 items
Supplier: Thermo Fisher Scientific
BATTERY KIT CORIOLIS COMPACT 1 * 1 items
Supplier: Bertin Instruments
MICROSWITCH ROLL.PLUNG MP320-0/375/200 1 * 1 items
Supplier: TUTTNAUER
Screw Cap Gl-10 1 * 1 items
Supplier: BUCHI
SAFETY VALVE 1 * 1 items
Supplier: CERTOCLAV
SHAFT KIT UHP 10 W/SEAL+ORING 1 * 1 KIT
Supplier: VWR Collection