Orders LinkContactUsLinkComponent
Objectives
Objectives
Objective LD A-plan, N.A. of 0,55, WD of 2,3 mm at cover glass 1,0
Catalog # ZEIS4212609906000
Supplier:   ZEISS
Objectives
Catalog # ZEIS4212609906000
Supplier:  ZEISS
Objectives
Objectives
Objective LD A-plan, N.A. of 0,55, WD of 2,3 mm at cover glass 1,0
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    40×
  • Description:
    Objective LD A-plan, N.A. of 0,55, WD of 2,3 mm at cover glass 1,0
  • Seq:
    15