Orders LinkContactUsLinkComponent
Objectives
Objectives
Objective LD A-plan, N.A. of 0,35, WD of 4,9 mm at cover glass 1,0
Catalog # ZEIS4212509901000
Supplier:   ZEISS
Objectives
Catalog # ZEIS4212509901000
Supplier:  ZEISS
Objectives
Objectives
Objective LD A-plan, N.A. of 0,35, WD of 4,9 mm at cover glass 1,0
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    20×
  • Description:
    Objective LD A-plan, N.A. of 0,35, WD of 4,9 mm at cover glass 1,0
  • Seq:
    13