,,2 components: 1000mg/l each of Si Ti in HNO3 5% HF 0.2% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,,,, 1 * 100 mL
Catalog # CMPLRM109672L1
Supplier: CUSTOM MADE CHEMICALS LAB