PRESSURE GAUGE 0-10 BAR 1 * 1 items
Catalog # WERN1701-032
Supplier: ENVIROFALK PHARMAWATERSYSTEMS
Frequently Bought Together
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Printer pape
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Accessories for PIPETBOY acu / pro
Supplier: Integra
HF Discharge Valve - Bibby 1 * 1 items
Supplier: Cole Parmer
Gas sensor 0-20% CO2 Steri max. 190 C 1 * 1 items
Supplier: MEMMERT
Protectionfilter 5/10ml 1 * 150 items
Supplier: EPPENDORF
DUCT TAPE ALUMINIUM 40MM/50M 1 * 1 items
Supplier: Binder
Syringe filters, SupaTop
Supplier: GILSON
Accessories for diaphragm pumps
Supplier: BUCHI
HOSE ASSY COMPLETE A4000 1 * 1 items
Supplier: Cole Parmer
O-RING 1 * 1 items
Supplier: IKA
CAROUSEL KIT, METAL FOR 10 BOTTLES Ø35MM 1 * 1 items
Supplier: UVP ULTRA VIOLET PRODUCTS
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach