AMPIVIEW HPV 16/18 RNA PROBES SET 1 * 2 mL
Catalog # ENZOENZGEN1462000
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Aluminium Foil Seals for Cold Storage
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
Pasteur pipettes, disposable
Supplier: Brand
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Centrifuge tubes, Falcon®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection
Serological pipettes
Supplier: Greiner Bio-One
Culture media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Serological Pipettes, Open End
Supplier: VWR Collection