Orders LinkContactUsLinkComponent
Phosphoramidon
Phosphoramidon
5 mg
Catalog # ENZOBMLPI1110005
Supplier:   Enzo Life Sciences
Phosphoramidon
Catalog # ENZOBMLPI1110005
Supplier:  Enzo Life Sciences
CAS Number:  
Phosphoramidon
Phosphoramidon
5 mg
CAS Number:  
Price
216,00 €
Log in for availability
1 * 5 mg
Product Details & Documents
Neprilysin inhibitor
Inhibits neutral endopeptidase 24,11 (NEP, neprilysin; EC 3.4.24.11) (Ki=2 nM), a cell surface zinc metalloprotease which cleaves bioactive peptides such as bradykinin, neurotensin, and enkephalin. Also inhibits neprilysin 2 (Ki~2 nM). Potent inhibitor of endothelin-converting enzyme (ECE) (IC50=0,68 μM).
Specifications
Specifications
  • Pk:
    5 mg
  • Cat. No.:
    ENZOBMLPI1110005
  • Protein/peptide name:
    Phosphoramidon