Orders LinkContactUsLinkComponent
REDOX library plates, SCREEN-WELL®
REDOX library plates, SCREEN-WELL®
Screen-Well™ REDOX library, 500 μl, 96
Catalog # ENZOBML28350500
Supplier:   Enzo Life Sciences
REDOX library plates, SCREEN-WELL®
Catalog # ENZOBML28350500
Supplier:  Enzo Life Sciences
REDOX library plates, SCREEN-WELL®
Screen-Well™ REDOX library, 500 μl, 96
Price
2 810,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
Screen-Well™ REDOX library contains 84 compounds with defined prooxidant or antioxidant activity.
A variety of structurally and mechanistically different compound classes are included such as hydroperoxides, polyphenolics, metal chelators, thiols, thiol traps, radical scavengers, lazaroids and glutathione modulators as well as small molecule enzyme mimetics for SOD and glutathione peroxidase. The library is a useful tool for studying the roles of pro- and antioxidant molecules in cells as well as for use in in vitro applications.
Compounds are dissolved in DMSO at 10 mM and aliquoted into deep-well plates.
Stable for one year from the date of receipt when stored at –80 °C and shipped on dry ice.
Specifications
  • No. of wells:
    96
  • Description:
    Screen-Well™ REDOX library
  • Seq:
    04
  • Well volume:
    500 μl
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.