Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
1 COMPONENT C8H5O4K 60MG/L IN H2O 1 * 500 mL
CMPLRM048417L5
:
CUSTOM MADE CHEMICALS LAB
1 COMPONENT C8H5O4K 60MG/L IN H2O 1 * 500 mL
CMPLRM048417L5
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
SAMPLE CHAMBER CLOSURE 1 * 12 items
: Epredia
Rose Bengal lactone 95%, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
1-Amino-4-bromoanthraquinone-2-sulphonic acid sodium salt, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Anti-GAPDH Rabbit Recombinant Multiclonal Antibody [clone: RM1050]
: Abcam
VARIO TOTAL CHLORINE DPD 25 ML 1 * 100 items
: Lovibond
REAGENT B FOR AMTAX 1 * 5.200 mL
: Hach
Potassium hydroxide ≥85%, flakes
: Thermo Fisher Scientific