COLUMN INSTALLATION FIXTURE ASSEMBLY 1 * 1 items
Catalog # CCDLNP-1501
Supplier: LUCIDITY
Frequently Bought Together
Short-term tubes for accuro®
Supplier: Dräger
Gas detection tube for formaldehyde
Supplier: Dräger
Accessories for Liquid Nitrogen Vessels, TR,TP and GT
Supplier: Air Liquide
Gas detection tube for hydrogen sulphide
Supplier: Dräger
UV lamps, EL series
Supplier: UVP ULTRA VIOLET PRODUCTS
CELL HOLDER FOR SPECTROPHOT. AQUANOVA 1 * 1 items
Supplier: Cole-Parmer
SP-9 Standard spindle 1 * 1 items
Supplier: IKA
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
pH/mV/°C meters, bench, SevenExcellence™ S400
Supplier: Mettler - Toledo
Accessories for measuring magnifiers
Supplier: ESCHENBACH OPTIK
Automated stainers, Cellstain® 6
Supplier: THARMAC GmbH
Includes power supply, USB cable and outlet adapters 1 * 1 items
Supplier: YSI ENVIRONMENTAL
Storage phosphor screens and cassettes
Supplier: Cytiva
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Supplier: VWR Collection
DK Series Digestion Units
Supplier: VELP SCIENTIFIC
Precellys® Evolution Touch Homogeniser
Supplier: Bertin Instruments