ContactUsLinkComponent
,,7 components: 100mg/l each of Sodium p-n-decylbenzenesulfonate [CAS:2627-06-7] Sodium dodecylbenzenesulfonate [CAS:25155-30-0] Sodium xylene sulfonate [CAS:1300-72-7] Sodium Decyl Sulfate [CAS:142-87-0] Sodium 1-undecyl sulfate [CAS:1072-24 1 * 5 Ampoul
  CMPLRD1064061.5
,,7 components: 100mg/l each of Sodium p-n-decylbenzenesulfonate [CAS:2627-06-7] Sodium dodecylbenzenesulfonate [CAS:25155-30-0] Sodium xylene sulfonate [CAS:1300-72-7] Sodium Decyl Sulfate [CAS:142-87-0] Sodium 1-undecyl sulfate [CAS:1072-24 1 * 5 Ampoul
  CMPLRD1064061.5
Frequently Bought Together