Orders LinkContactUsLinkComponent
VWR® Filter Vials
VWR® Filter Vials
Nylon, 0,20 µm
Catalog # 548-9044A
Supplier:   VWR Collection
VWR® Filter Vials
Catalog # 548-9044A
Supplier:  VWR Collection
VWR® Filter Vials
Nylon, 0,20 µm
Price
247,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Filter vials with membrane, PE snap ring cap (11 mm), with hole and silicone white/PTFE red septa.
Vials have different colour caps for different membranes: RC with grey caps, PVDF with yellow caps, Nylon with light-blue caps, PTFE with pink caps and PES with light-green caps.
Specifications
  • Membrane:
    Nylon
  • Pore size:
    0,20 µm
  • Seq:
    01