Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TISSU PROFITEXTRASMOOTH 55CELL 45POL 68 1 * 5 Roll
Catalog # ECOLPTS0060R01500W
Supplier:
ECOLINE NV SA
Expand image
TISSU PROFITEXTRASMOOTH 55CELL 45POL 68 1 * 5 Roll
Catalog # ECOLPTS0060R01500W
Supplier:
ECOLINE NV SA
Frequently Bought Together
Reaction block with 8 undivided cells 1 * 1 items
Supplier: IKA
Cleaning cloths, J-Cloth® Plus
Supplier: PGI NONWOVEN
SENSOLYT 700 IQ/SET 1 * 1 items
Supplier: WTW ONLINE
PROSAT® QuickTask™ Mopping System
Supplier: CONTEC