Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
26 COMPONENTS: 100UG/ML EACH 1 * 1 mL
Catalog # ACSDF263511
Supplier:
CPACHEM
Expand image
26 COMPONENTS: 100UG/ML EACH 1 * 1 mL
Catalog # ACSDF263511
Supplier:
CPACHEM
Frequently Bought Together
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Reagents for DR800, DR900, DR2010, DR2500, DR2400, DR2800, DR3800, DR3900, DR5000, DR6000
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach