Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1 COMPONENT NITRATES 100 MG/L IN WATER 1 * 100 mL
Catalog # CMPLRM000583L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
1 COMPONENT NITRATES 100 MG/L IN WATER 1 * 100 mL
Catalog # CMPLRM000583L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Sodium hydroxide 32% in aqueous solution
Supplier: Roth Carl
Fast red ITR base, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
20G PL SILICA HR 1 (142 TESTS) 1 * 20 g
Supplier: WATER ID PRIMELAB
Anti-Cdc25A (phospho S18) Rabbit Recombinant Multiclonal Antibody [clone: RP23040112]
Supplier: Abcam