VWR® Viscometers
Calibrated, size: 0B, 1 to 5 cSt, 0,005 cSt/s K
Catalog # VWRU10791-458
Supplier: VWR Collection
Jump to:
Product Details & Documents
The Cannon-Fenske routine viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104.
Minimum sample volume is 11 ml. Minimum bath depth is 241 mm (9,5")
Specifications
- Range:1 to 5 cSt
- Description:Calibrated, size: 0B
- Constant:0,005 cSt/s K