Orders LinkContactUsLinkComponent
VWR® Viscometers
VWR® Viscometers
Calibrated, size: 0C, 0,6 to 3 cSt, 0,003 cSt/s K
Catalog # VWRU10791-456
Supplier:   VWR Collection
VWR® Viscometers
Catalog # VWRU10791-456
Supplier:  VWR Collection
VWR® Viscometers
VWR® Viscometers
Calibrated, size: 0C, 0,6 to 3 cSt, 0,003 cSt/s K
Price
432,00 €
Log in for availability
1 * 1 items
Product Details & Documents
The Cannon-Fenske routine viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104.
Minimum sample volume is 11 ml. Minimum bath depth is 241 mm (9,5")
Specifications
  • Range:
    0,6 to 3 cSt
  • Description:
    Calibrated, size: 0C
  • Constant:
    0,003 cSt/s K
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.