VWR® Viscometers
Calibrated, size: 0C, 0,6 to 3 cSt, 0,003 cSt/s K
Catalog # VWRU10791-456
Supplier: VWR Collection
Jump to:
Product Details & Documents
The Cannon-Fenske routine viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104.
Minimum sample volume is 11 ml. Minimum bath depth is 241 mm (9,5")
Specifications
- Range:0,6 to 3 cSt
- Description:Calibrated, size: 0C
- Constant:0,003 cSt/s K