Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MULTI-ELEMEN 27 COMPONENTS: 1000MG/L 1 * 100 mL
Catalog # CMPLRM102688L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
MULTI-ELEMEN 27 COMPONENTS: 1000MG/L 1 * 100 mL
Catalog # CMPLRM102688L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
pH buffer solutions, colour coded
Supplier: Hach
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond