ContactUsLinkComponent
Y-27632 is selective and potent ROCK inhibitor. Y-27632 plays various roles including contraction of vascular smooth muscle cells vi a ROCK signaling pathways. Furthermore, it suppresses the cell death of human embryonic stem cells (hESCs) and human 1 * 5 mg
  WAKF259-00613
Y-27632 is selective and potent ROCK inhibitor. Y-27632 plays various roles including contraction of vascular smooth muscle cells vi a ROCK signaling pathways. Furthermore, it suppresses the cell death of human embryonic stem cells (hESCs) and human 1 * 5 mg
  WAKF259-00613
Frequently Bought Together