ContactUsLinkComponent
CTP is Cytidine-5'-triphosphate, the Chinese name is cytidine triphosphate, which is an extremely stable nucleotide. This compound a nd its sodium salt are essential components of energy conversion in biological systems and can be used in a variety o 1 * 20 mL
Catalog # ABNOU0512.20ML
Supplier:  Abnova
CTP is Cytidine-5'-triphosphate, the Chinese name is cytidine triphosphate, which is an extremely stable nucleotide. This compound a nd its sodium salt are essential components of energy conversion in biological systems and can be used in a variety o 1 * 20 mL
Catalog # ABNOU0512.20ML
Supplier:  Abnova
Frequently Bought Together