ContactUsLinkComponent
UTP is Uridine-5'-triphosphate, which is an extremely stable nucleotide. This compound and its sodium salt are essential components of energy conversion in biological systems and can be used in various related applications in molecular biology, such 1 * 1 mL
Catalog # ABNOU0515.1ML
Supplier:  Abnova
UTP is Uridine-5'-triphosphate, which is an extremely stable nucleotide. This compound and its sodium salt are essential components of energy conversion in biological systems and can be used in various related applications in molecular biology, such 1 * 1 mL
Catalog # ABNOU0515.1ML
Supplier:  Abnova
Frequently Bought Together