ContactUsLinkComponent
Murine RNase inhibitor binds to RNase A, B or C in a 1:1 ratio through non-covalent bonding, thereby inhibiting the activity of the enzymes and protecting RNA from degradation. This product with activity data is belong to bioactive protein. 1 * 1 mL
  ABNOU0440.1ML
 :  Abnova
Murine RNase inhibitor binds to RNase A, B or C in a 1:1 ratio through non-covalent bonding, thereby inhibiting the activity of the enzymes and protecting RNA from degradation. This product with activity data is belong to bioactive protein. 1 * 1 mL
  ABNOU0440.1ML
 :  Abnova
Frequently Bought Together